Learn More
Cayman Chemical Tiotropium (bromide), Purity: ≥98%, CAS: 136310-93-5, 50 mg
SDP
Catalog No. 502972451
Encompass_Preferred
An antagonist that binds to M1, M2, and M3 muscarinic acetylcholine receptors (Kds = 0.43, 0.54, and 0.69 nM, respectively, for human receptors); decreases acetylcholine-induced contraction of isolated guinea pig trachea in a concentration-dependent manner; confers complete protection against acetylcholine-induced bronchospasms in anesthetized dogs at 1.0 g/L inhaled aerosol
Catalog No. 50-297-2451 Supplier Cayman Chemical Supplier No. 1577350 MG
An antagonist that binds to M1, M2, and M3 muscarinic acetylcholine receptors (Kds = 0.43, 0.54, and 0.69 nM, respectively, for human receptors); decreases acetylcholine-induced contraction of isolated guinea pig trachea in a concentration-dependent manner; confers complete protection against acetylcholine-induced bronchospasms in anesthetized dogs at 1.0 g/L inhaled aerosol