Learn More
Apexbio Technology LLC Tioxolone 4991-65-5 1g
SDP
Catalog No. 502240417
Encompass_Preferred
Tioxolone (CAS 4991-65-5) is a small molecule inhibitor that targets metalloenzyme carbonic anhydrase I binding with high affinity as indicated by a Ki value of 91 nM By interfering with the catalytic activity of carbonic anhydrase I tioxolone modulates processes dependent on rapid CO hydration which is relevant in physiological and pathological contexts involving pH regulation and gas exchange This compound is utilized in biomedical research to study the functional role of carbonic anhydrase I and to explore potential therapeutic strategies involving the inhibition of metalloenzyme activity
This item is not returnable. View return policy
Catalog No. 50-224-0417 Supplier Apexbio Technology LLC Supplier No. A43621000
Tioxolone (CAS 4991-65-5) is a small molecule inhibitor that targets metalloenzyme carbonic anhydrase I binding with high affinity as indicated by a Ki value of 91 nM By interfering with the catalytic activity of carbonic anhydrase I tioxolone modulates processes dependent on rapid CO hydration which is relevant in physiological and pathological contexts involving pH regulation and gas exchange This compound is utilized in biomedical research to study the functional role of carbonic anhydrase I and to explore potential therapeutic strategies involving the inhibition of metalloenzyme activity