Learn More
Pion Inc Pion Inc. 1 mm Pathlength Right-Hand Tip, Precise 1 mm Optical Pathlength, Real-Time Concentration Measurements, Interchangeable
Catalog No. NC2744775
Encompass
The Pion Inc. 1 mm Pathlength Right-Hand Tip is designed for advanced spectroscopic analysis, enhancing real-time concentration measurements in small-volume dissolution studies. This interchangeable accessory is compatible with dip-style fiber optic probes and integrates seamlessly with Pion's MicroDISS and MiniDISS Profiler systems, ensuring high-density data collection for evaluating drug formulations effectively.
Catalog No. NC2744775 Supplier Pion Inc Supplier No. 112292
The Pion Inc. 1 mm Pathlength Right-Hand Tip is designed for advanced spectroscopic analysis, enhancing real-time concentration measurements in small-volume dissolution studies. This interchangeable accessory is compatible with dip-style fiber optic probes and integrates seamlessly with Pion's MicroDISS and MiniDISS Profiler systems, ensuring high-density data collection for evaluating drug formulations effectively.