Learn More
Sarstedt Inc Pipette tip, 1000 µl, transparent, PCR Performance Tested, 96/pk
Catalog No. NC1896076 Shop All Sarstedt Inc. Products
Encompass_Preferred
Pipette tip, working volume: 1000 µl, transparent, filling level rings, PCR Performance Tested, suitable for Eppendorf, Gilson, Finnpipette, Biohit, Brand and Socorex and products of identical design, 96 piece(s)/box.
Catalog No. NC1896076 Supplier Sarstedt Inc Supplier No. 70.3050.200
Pipette tip, working volume: 1000 µl, transparent, filling level rings, PCR Performance Tested, suitable for Eppendorf, Gilson, Finnpipette, Biohit, Brand and Socorex and products of identical design, 96 piece(s)/box.