Learn More
Labcon North America Eclipse™ 1000uL Blue Pipet Tips, in Resealable Bags
Catalog No. 50101629
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Blue color; 1000 tips per bag; 10 bags per case; 75.9mm. H; Made in USA
Catalog No. 50-101-629 Supplier Labcon North America Supplier No. 10418000009
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Blue color; 1000 tips per bag; 10 bags per case; 75.9mm. H; Made in USA