Learn More
Labcon North America Eclipse™ 1000uL Blue Pipet Tips, in 96 Stack Rack
Catalog No. 50101630
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Blue color; 96 tips per rack; 5 racks per stack ; 10 stacks per case; 75.9mm. H; Made in USA
Catalog No. 50-101-630 Supplier Labcon North America Supplier No. 10417000009
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Blue color; 96 tips per rack; 5 racks per stack ; 10 stacks per case; 75.9mm. H; Made in USA