Learn More
Labcon North America Eclipse™ 1000uL Blue Pipet Tips, in 96 Racks, Sterile
Catalog No. 50101632
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Blue color; 96 tips per rack; 6 racks per pack; 8 packs per case; 75.9mm. H; Made in USA
Catalog No. 50-101-632 Supplier Labcon North America Supplier No. 10419650089
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Blue color; 96 tips per rack; 6 racks per pack; 8 packs per case; 75.9mm. H; Made in USA