Learn More
Labcon North America Eclipse™ 1000uL Blue Pipet Tips, in 96 Racks
Catalog No. 50101631
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; ; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Blue color; 96 tips per rack; 6 racks per pack; 8 packs per case; 75.9mm. H; Made in USA
Catalog No. 50-101-631 Supplier Labcon North America Supplier No. 10419600089
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 1000uL Pipettors ; UltraFine™ Point; ; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Blue color; 96 tips per rack; 6 racks per pack; 8 packs per case; 75.9mm. H; Made in USA