Learn More
Tecan 10L Tips ANSI NFil Ref
Catalog No. 501305997
Encompass_Preferred
Genuine Tecan 10 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, non-filtered, conductive. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. These tips are designed to fit into a Disposable Tip (DiTi) boxas a refill. 24 x 96-tip racks, 2,304 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials.
This item is not returnable. View return policy
Catalog No. 50-130-5997 Supplier Tecan Supplier No. 30104973
Genuine Tecan 10 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, non-filtered, conductive. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. These tips are designed to fit into a Disposable Tip (DiTi) boxas a refill. 24 x 96-tip racks, 2,304 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials.