Learn More
Labcon North America Eclipse™ 10uL Graduated Pipet Tips with UltraFine™ Point, in Resealable Bags
Catalog No. 50101636
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ ; Graduated at 2ul and 10uL; With UltraFine™ point; Fits popular 10uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 38.1mm. H; Made in USA
Catalog No. 50-101-636 Supplier Labcon North America Supplier No. 10388000009
Tip, Pipet; Labcon; Eclipse™ ; Graduated at 2ul and 10uL; With UltraFine™ point; Fits popular 10uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 38.1mm. H; Made in USA