Learn More
Labcon North America Eclipse™ 10uL Graduated Pipet Tips with UltraFine™ Point, in 96 Racks
Catalog No. 50101637
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ ; Graduated at 2uL and 10uL ; With UltraFine™ point; Fits popular 10uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; 96 tips/rack; 12 racks per pack; 10 packs per case; 31.4mm. H; Made in USA
Catalog No. 50-101-637 Supplier Labcon North America Supplier No. 10389600389
Tip, Pipet; Labcon; Eclipse™ ; Graduated at 2uL and 10uL ; With UltraFine™ point; Fits popular 10uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; 96 tips/rack; 12 racks per pack; 10 packs per case; 31.4mm. H; Made in USA