Learn More
Labcon North America Eclipse™ 10uL Graduated Pipet Tips with UltraFine™ Point, in 96 Racks, Sterile
Catalog No. 50101638
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ ; Graduated at 2uL and 10uL ; With UltraFine™ point; Fits popular 10uL pipettors; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 12 racks per pack; 10 packs per case; 31.4mm. H; Made in USA
Catalog No. 50-101-638 Supplier Labcon North America Supplier No. 10389650389
Tip, Pipet; Labcon; Eclipse™ ; Graduated at 2uL and 10uL ; With UltraFine™ point; Fits popular 10uL pipettors; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 12 racks per pack; 10 packs per case; 31.4mm. H; Made in USA