Learn More
Thomas Scientific TIP, 1250UL, RACKED, STERILE, 6X96
SDP
Catalog No. 501460578
Encompass
These tips are the latest in low retention polymer technology and come to an extra fine point at the dispensing orifice, minimizing inaccuracies caused by surface adhesion and droplet retention. Their universal fit sealing ring design forms a superior seal on a broad range of pipette brands, including hard-to-fit P-20 pipettes. The 1000µl size are larger and more finely tapered (compared to traditional tips), allowing them to reach bottoms of 12 x 75mm culture tubes, 0.5ml tubes and even 0.2ml PCR tubes. All sizes are clear and metal free.
Catalog No. 50-146-0578 Supplier Thomas Scientific Supplier No. 1158U40
These tips are the latest in low retention polymer technology and come to an extra fine point at the dispensing orifice, minimizing inaccuracies caused by surface adhesion and droplet retention. Their universal fit sealing ring design forms a superior seal on a broad range of pipette brands, including hard-to-fit P-20 pipettes. The 1000µl size are larger and more finely tapered (compared to traditional tips), allowing them to reach bottoms of 12 x 75mm culture tubes, 0.5ml tubes and even 0.2ml PCR tubes. All sizes are clear and metal free.