Learn More
Roboz Surgical TIP .17X.10MM PATTERN #3 120MM
SDP
Catalog No. NC9001227
Encompass
Dumont Tweezers; Pattern #3; .17 X .10mm Tip Size; 120mm Length; Material: Dumostar
Catalog No. NC9001227 Supplier Roboz Surgical Supplier No. RS4964
Dumont Tweezers; Pattern #3; .17 X .10mm Tip Size; 120mm Length; Material: Dumostar