Learn More
Thomas Scientific PCR 8-Strips, 0.15mL volume/tube, individually attached Flat Caps, Clear; Suitable for qPCR; Rigid
SDP
Catalog No. 501460494
Encompass_Preferred
BRAND PCR tube strips are manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications. These tube strips are manufactured without lubricants or chemical additives that can contaminate reagents and samples. PCR tubes and plates from BRAND are compatible with most 24-, 32-, 96-, and 384-well thermal blocks. BRAND PCR tubes and plates are autoclavable and RNase-, DNase-, and DNA-free. NEW! PCR 8-strips are now available in convenient tube/cap sets. The tube strips feature thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.
Catalog No. 50-146-0494 Supplier Thomas Scientific Supplier No. 1162F07
BRAND PCR tube strips are manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications. These tube strips are manufactured without lubricants or chemical additives that can contaminate reagents and samples. PCR tubes and plates from BRAND are compatible with most 24-, 32-, 96-, and 384-well thermal blocks. BRAND PCR tubes and plates are autoclavable and RNase-, DNase-, and DNA-free. NEW! PCR 8-strips are now available in convenient tube/cap sets. The tube strips feature thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.