Learn More
Labcon North America Eclipse™ FlexTop™ 300uL Extended Pipet Tips with UltraFine™ points, in 96 Racks
Catalog No. 50101705
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extra long; Fits popular 300uLpipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks per pack; 8 packs per case; 71.1mm. H; Made in USA
Catalog No. 50-101-705 Supplier Labcon North America Supplier No. 10149600089
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extra long; Fits popular 300uLpipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks per pack; 8 packs per case; 71.1mm. H; Made in USA