Learn More
Medchemexpress LLC TIP 39, Tuberoinfundibular Neuropeptide | 277302-47-3 | 99.84% | 4504.20 | 1 MG
Supplier: Medchemexpress LLC HYP18521MG
TIP 39, Tuberoinfundibular Neuropeptide is an endogenous PTH2 receptor agonist and antihypertensive agent. It selectively activates the PTH2 receptor without activity on the PTH1 receptor, stimulating cAMP production, activating adenylate cyclase, and elevating intracellular calcium levels from intracellular stores. This neuropeptide is highly conserved in humans, mice, and rats.
- Selectively activates PTH2 receptor with no activity on PTH1 receptor
- Stimulates cAMP production
- Activates adenylate cyclase
- Elevates intracellular calcium levels via mobilization from intracellular stores
- Highly conserved across humans, mice, and rats
- Applicable to research related to nociception and inflammation-induced pain