Learn More
Labcon North America ZAP™ 100uL Graduated Aerosol Filter Pipet Tips, in Racks, Sterile
Catalog No. 50101591
Encompass_Preferred
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; With Graduations; Fit most popular 100uL Pipettors; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 10 racks/pack; 10 packs/case; 49.7mm H.; Made in USA
Catalog No. 50-101-591 Supplier Labcon North America Supplier No. 10929650089
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; With Graduations; Fit most popular 100uL Pipettors; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 10 racks/pack; 10 packs/case; 49.7mm H.; Made in USA