Learn More
Labcon North America ZAP™ 1000uL Wide Orifice Aerosol Filter Pipet Tips, in Racks, Sterile
Catalog No. 50101594
Encompass_Preferred
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; With wide orifice; Fits most popular 1000uL Pipettors; Sterile; With 2.03mm wide orifice for viscous samples; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 6 racks/pack; 8 packs/case; 88.9mm H.; Made in USA
Catalog No. 50-101-594 Supplier Labcon North America Supplier No. 11999650089
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; With wide orifice; Fits most popular 1000uL Pipettors; Sterile; With 2.03mm wide orifice for viscous samples; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 6 racks/pack; 8 packs/case; 88.9mm H.; Made in USA