Learn More
Labcon North America ZAP™ 10uL Aerosol Filter Pipet Tips with TubeGard™, in Racks, Sterile
Catalog No. 50101583
Encompass_Preferred
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; With TubeGard™; Fits most popular 10uL Pipettors; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 10 racks/pack; 10 packs/case; 38.1mm H.; Made in USA
Catalog No. 50-101-583 Supplier Labcon North America Supplier No. 11519650089
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; With TubeGard™; Fits most popular 10uL Pipettors; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 10 racks/pack; 10 packs/case; 38.1mm H.; Made in USA