Learn More
Nordson EFD 45 Bend, 12.7 mm, 0.5"" Angled Tips, Blue, 50 per box
SDP
Catalog No. NC1557804
Encompass
This item has a minimum qty of 2 per supplier requirements.
Optimum angled stainless steel tips with SafetyLok™ hubs deliver accurate, repeatable fluid dispensing in challenging applications. Each tip has a precise bend radius, resulting in better consistency, higher productivity, and decreased waste when dispensing in hard-to-reach areas. The unique SafetyLok hub thread ensures safe, secure attachment to the syringe barrel. Each hub is designed with flush mating of the hub to the cutoff for smooth fluid flow and to prevent trapped air, which causes dripping and drooling. Burr-free passivated stainless steel tubing ensures consistent fluid deposits.
Catalog No. NC1557804 Supplier Nordson EFD Supplier No. 7018273
This item has a minimum qty of 2 per supplier requirements.
Optimum angled stainless steel tips with SafetyLok™ hubs deliver accurate, repeatable fluid dispensing in challenging applications. Each tip has a precise bend radius, resulting in better consistency, higher productivity, and decreased waste when dispensing in hard-to-reach areas. The unique SafetyLok hub thread ensures safe, secure attachment to the syringe barrel. Each hub is designed with flush mating of the hub to the cutoff for smooth fluid flow and to prevent trapped air, which causes dripping and drooling. Burr-free passivated stainless steel tubing ensures consistent fluid deposits.