Learn More
Health Products For You Becton Dickinson Syringe Tip Cap, Black, Sterile, 10/Pack
SDP
Catalog No. NC3781399
Encompass_Preferred
Becton Dickinson Syringe Tip Cap offers a slip-on design that allows a quick application, and self-venting features reduce additional manipulation. The cover peels back have access tip caps as needed, preserving sterility for the remaining caps. The tip cap is also designed with sealed tray sections, saving time and costs while maintaining sterility.
This item is not returnable. View return policy
Catalog No. NC3781399 Supplier Health Products For You Supplier No. 308341PK
Becton Dickinson Syringe Tip Cap offers a slip-on design that allows a quick application, and self-venting features reduce additional manipulation. The cover peels back have access tip caps as needed, preserving sterility for the remaining caps. The tip cap is also designed with sealed tray sections, saving time and costs while maintaining sterility.