Learn More
Genesee Scientific Corporation 20 ul Ergo X Barrier Tip, 960 tips/cs
SDP
Catalog No. NC1842408
Encompass
Low retention polymer maximizes sample recovery and improves accuracy. A positive stop that inhibits jamming and eliminates high ejection forces. creates a secure seal with less insertion force than competing tips.
Catalog No. NC1842408 Supplier Genesee Scientific Corporation Supplier No. 24701
Low retention polymer maximizes sample recovery and improves accuracy. A positive stop that inhibits jamming and eliminates high ejection forces. creates a secure seal with less insertion force than competing tips.