Learn More
Labcon North America Eclipse™ 50uL Pipet Tips for 16 Channel Finnpipette Pipettors, in 384 Rack, Sterile
Catalog No. 50101683
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; Fits 16 channel Finnpipette; Sterile; ATP, Rnase/Dnase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 384 tips per rack; 10 racks per pack; 5 packs per case; 32.9mm. H; Made in USA
Catalog No. 50-101-683 Supplier Labcon North America Supplier No. 10076950089
Tip, Pipet; Labcon; Eclipse™; Specialty; Fits 16 channel Finnpipette; Sterile; ATP, Rnase/Dnase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 384 tips per rack; 10 racks per pack; 5 packs per case; 32.9mm. H; Made in USA