Learn More
Labcon North America Eclipse™ 50uL Pipet Tips for 16 Channel Finnpipette Pipettors, in 384 Rack
Catalog No. 50101682
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; Fits 16 channel Finnpipette; ATP, Rnase/Dnase and Endotoxin Free (non-pyrogenic); polypropylene USP Classs IV resin; Autoclavable; 384 tips per rack; 10 racks per pack; 5 packs per case; 32.9mm. H; Made in USA
Catalog No. 50-101-682 Supplier Labcon North America Supplier No. 10076900089
Tip, Pipet; Labcon; Eclipse™; Specialty; Fits 16 channel Finnpipette; ATP, Rnase/Dnase and Endotoxin Free (non-pyrogenic); polypropylene USP Classs IV resin; Autoclavable; 384 tips per rack; 10 racks per pack; 5 packs per case; 32.9mm. H; Made in USA