Learn More
Labcon North America Eclipse™ 200uL Gel Loading Capillary Pipet Tips, in Resealable Bags
Catalog No. 50101707
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; With Gel Loading Capillary; Fits popular 200uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 82.5mm. H; Made in USA
Catalog No. 50-101-707 Supplier Labcon North America Supplier No. 10348000009
Tip, Pipet; Labcon; Eclipse™; With Gel Loading Capillary; Fits popular 200uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 82.5mm. H; Made in USA