Learn More
Labcon North America Eclipse™ 200uL Wide Orifice Pipet Tips, in Resealable Bags
Catalog No. 50101666
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; Wide orifice; Fits popular 200uL pipettors; With 1.45mm orifice for viscous samples; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips per bag; 10 bags per case; 49.7mm. H; Made in USA
Catalog No. 50-101-666 Supplier Labcon North America Supplier No. 10268000009
Tip, Pipet; Labcon; Eclipse™; Specialty; Wide orifice; Fits popular 200uL pipettors; With 1.45mm orifice for viscous samples; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips per bag; 10 bags per case; 49.7mm. H; Made in USA