Learn More
Labcon North America Eclipse™ 200uL Wide Orifice Pipet Tips, in Racks, Sterile
Catalog No. 50101668
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; Wide orifice; Fits popular 200uL pipettors; With 1.45mm orifice for viscous samples; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips per rack; 10 racks per pack; 10 packs per case; 49.7mm. H; Made in USA
Catalog No. 50-101-668 Supplier Labcon North America Supplier No. 10269650089
Tip, Pipet; Labcon; Eclipse™; Specialty; Wide orifice; Fits popular 200uL pipettors; With 1.45mm orifice for viscous samples; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips per rack; 10 racks per pack; 10 packs per case; 49.7mm. H; Made in USA