Learn More
Perkin Elmer US LLC GemTip Kit for Cross-Flow II Nebulizer
Catalog No. 509052272
Encompass_Preferred
These replacement tips are included in the ELAN DRC-e/9000 Spares Kit (N8120610). Note: For use with the Cross-Flow II Nebulizers only.
Catalog No. 50-905-2272 Supplier Perkin Elmer US LLC Supplier No. N8120515
These replacement tips are included in the ELAN DRC-e/9000 Spares Kit (N8120610). Note: For use with the Cross-Flow II Nebulizers only.