Learn More
Labcon North America Eclipse™ 1200uL Pipet Tips for Rainin LTS Pipettors, in Resealable Bags
Catalog No. 50101648
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; 1200uL; Fits 1200uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips per bag; 10 bags per case; 88.9mm. H; Made in USA
Catalog No. 50-101-648 Supplier Labcon North America Supplier No. 10198000009
Tip, Pipet; Labcon; Eclipse™; Specialty; 1200uL; Fits 1200uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips per bag; 10 bags per case; 88.9mm. H; Made in USA