Learn More
Labcon North America Eclipse™ 1200uL Pipet Tips for Rainin LTS Pipettors, in 96 Racks, Sterile
Supplier:  Labcon North America 10199650089
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; 1200uL; Fits 1200uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 6 racks/pk.; 8 pk./case; 88.9mm; Made in USA
Catalog No. 50-101-650