Learn More
Labcon North America Eclipse™ 20uL Pipet Tips for Rainin LTS Pipettors, in 96 Racks, Sterile
Catalog No. 50101671
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; 20uL; Fits 20uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 960 tips per pack; 10 packs per case; 48.3mm. H; Made in USA
Catalog No. 50-101-671 Supplier Labcon North America Supplier No. 10119650089
Tip, Pipet; Labcon; Eclipse™; Specialty; 20uL; Fits 20uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 960 tips per pack; 10 packs per case; 48.3mm. H; Made in USA