Learn More
Labcon North America Eclipse™ 250uL Pipet Tips for Rainin LTS Pipettors, in Resealable Bags
Catalog No. 50101672
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; 250uL; Fits 250uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips per bag; 10 bags per case; 51.3mm. H; Made in USA
Catalog No. 50-101-672 Supplier Labcon North America Supplier No. 10188000009
Tip, Pipet; Labcon; Eclipse™; Specialty; 250uL; Fits 250uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips per bag; 10 bags per case; 51.3mm. H; Made in USA