Learn More
Labcon North America Eclipse™ Macro 5mL Pipet Tips for Popular Pipettors, in Resealable Bags
Catalog No. 50101661
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; For Popular Macro pipettors; 1-5mL; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 250/bag; 10 bags/case; 144.3mm H.; Made in USA
Catalog No. 50-101-661 Supplier Labcon North America Supplier No. 10288600009
Tip, Pipet; Labcon; Eclipse™; For Popular Macro pipettors; 1-5mL; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 250/bag; 10 bags/case; 144.3mm H.; Made in USA