Learn More
Labcon North America Eclipse™ Macro 10mL Pipet Tips for Popular Pipettors, in Resealable Bags
Catalog No. 50101684
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; For Popular Macro pipettors; 5-10mL; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 250/bag; 10 bags/case; 155.7mm H.; Made in USA
Catalog No. 50-101-684 Supplier Labcon North America Supplier No. 10298600009
Tip, Pipet; Labcon; Eclipse™; For Popular Macro pipettors; 5-10mL; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 250/bag; 10 bags/case; 155.7mm H.; Made in USA