Learn More
Labcon North America Eclipse™ 1250uL Extended Length Pipet Tips for Matrix Pipettors, in Resealable Bags
Catalog No. 50101657
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Extended Length; Fits Matrix 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 88.9mm. H; Made in USA
Catalog No. 50-101-657 Supplier Labcon North America Supplier No. 10478000009
Tip, Pipet; Labcon; Eclipse™; Extended Length; Fits Matrix 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 88.9mm. H; Made in USA