Learn More
Labcon North America Eclipse™ 1250uL Extended Length Pipet Tips for Matrix Pipettors, in Racks
Catalog No. 50101658
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Extended Length; Fits Matrix 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks/pack; 8 packs per case; 88.9mm. H; Made in USA
Catalog No. 50-101-658 Supplier Labcon North America Supplier No. 10479600089
Tip, Pipet; Labcon; Eclipse™; Extended Length; Fits Matrix 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks/pack; 8 packs per case; 88.9mm. H; Made in USA