Learn More
Labcon North America Eclipse™ 10uL Pipet Tips with Tubegard™, in 96 Racks, Sterile
Catalog No. 50101653
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ ; Extended Length with TubeGard™; Fits popular 10uL pipettors; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 38.1mm. H; Made in USA
Catalog No. 50-101-653 Supplier Labcon North America Supplier No. 10369650089
Tip, Pipet; Labcon; Eclipse™ ; Extended Length with TubeGard™; Fits popular 10uL pipettors; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 38.1mm. H; Made in USA