Learn More
Labcon North America Eclipse™ FlexTop™ 30uL Pipet Tips with UltraFine™ points, in 96 Racks, Sterile
Catalog No. 50101681
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Fits popular 10uL pipettors; Low mounting and ejection forces less than 3kg; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 38.1mm. H; Made in USA
Catalog No. 50-101-681 Supplier Labcon North America Supplier No. 10169650089
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Fits popular 10uL pipettors; Low mounting and ejection forces less than 3kg; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 38.1mm. H; Made in USA