Learn More
Andwin Scientific Universal Fit Tips, 200 uL, Clear, Pre-sterile, 960/pk
SDP
Catalog No. NC1220123
Encompass_Preferred
These universal pipet tips are made from virgin polypropylene. The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration that reduces the carbon footprint.
Catalog No. NC1220123 Supplier Andwin Scientific Supplier No. 2102NS
These universal pipet tips are made from virgin polypropylene. The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration that reduces the carbon footprint.