Learn More
Andwin Scientific Neptune Universal Pipette Tips, Universal Fit Tips, 200 µL, Clear, 96/Rack
SDP
Catalog No. NC0894960
Encompass_Preferred
The Neptune Non-filt Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 200 L. This product is sold in a pack of 960 tips.
Catalog No. NC0894960 Supplier Andwin Scientific Supplier No. 89140896
The Neptune Non-filt Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 200 L. This product is sold in a pack of 960 tips.