Learn More
Andwin Scientific OneTouch Pipet Tip, 200 uL, Non-Sterile, 4800/cs
SDP
Catalog No. 501060574
Encompass_Preferred
Featuring a dual-shot tip design, these pipet tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft, easy-to-seal, color-coded collar. The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Catalog No. 501060574 Supplier Andwin Scientific Supplier No. 10330
Featuring a dual-shot tip design, these pipet tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft, easy-to-seal, color-coded collar. The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.