Learn More
Andwin Scientific TIP ONETOUCH 200 STERILE 960PK
SDP
Supplier:  Andwin Scientific 10340
Encompass_Preferred
The sterile OneTouch Pipet Tips feature accuracy and high-quality performance. The product is compatible with a huge variety of pipettors and is free of RNase, DNase, and pyrogens. The OneTouch tips provide a leak proof seal and require a minimal force to be ejected. Tips have a volume capacity of 200 L (1330 L Range). This product is packaged in plastic racks and is sold in a pack of 960.
Catalog No. NC0318512