Learn More
Molecular Devices Molecular Devices Pipette Tips, 384-Well, Black, 30 μL
Catalog No. 5000014381
Encompass_Preferred
Disposable 384-well pipette tips designed for FLIPR and FlexStation 3 fluidics modules. These black, 30 μL tips are supplied in 384-tip racks and packaged 50 racks per case. They are intended for automated 16-channel pipettor heads to minimize cross-contamination in high-throughput assays.
  • 384-well rack format.
  • 30 μL nominal capacity.
  • Black color.
  • Non-sterile.
  • Packaged 50 racks per case.
  • Designed for use with 16-channel pipettor heads.
Catalog No. 50-000-14381 Supplier Molecular Devices Supplier No. 90000764
Disposable 384-well pipette tips designed for FLIPR and FlexStation 3 fluidics modules. These black, 30 μL tips are supplied in 384-tip racks and packaged 50 racks per case. They are intended for automated 16-channel pipettor heads to minimize cross-contamination in high-throughput assays.
  • 384-well rack format.
  • 30 μL nominal capacity.
  • Black color.
  • Non-sterile.
  • Packaged 50 racks per case.
  • Designed for use with 16-channel pipettor heads.