Learn More
Molecular Devices Molecular Devices Pipette Tips, 384-Well, Clear, 30 μL
Catalog No. 5000014398
Encompass_Preferred
Molecular Devices FLIPR pipette tips are precision consumables designed for reliable liquid handling in high-throughput cellular screening applications. These tips are manufactured under controlled conditions to ensure consistent fit and performance with tip blocks used in plate-based assays.
  • 384-well format for high-density microplate workflows.
  • Clear tips optimized for optical assays and visibility.
  • Nominal capacity of 30 μL per tip.
  • Certified RNase-/DNase-free and nonpyrogenic; nonsterile.
  • Packaged as multiple racks, 50 racks per case for bulk use.
Catalog No. 50-000-14398 Supplier Molecular Devices Supplier No. 90000763
Molecular Devices FLIPR pipette tips are precision consumables designed for reliable liquid handling in high-throughput cellular screening applications. These tips are manufactured under controlled conditions to ensure consistent fit and performance with tip blocks used in plate-based assays.
  • 384-well format for high-density microplate workflows.
  • Clear tips optimized for optical assays and visibility.
  • Nominal capacity of 30 μL per tip.
  • Certified RNase-/DNase-free and nonpyrogenic; nonsterile.
  • Packaged as multiple racks, 50 racks per case for bulk use.