Learn More
CELLTREAT Scientific Products 10µL Extended Length Pipette Tip Rack, Empty Rack for LfTS Reload System, Non-Sterile
SDP
Catalog No. NC3377676
Encompass_Preferred
Empty LfTS Tip Racks available without a wafer to facilitate refills with LfTS Reload SystemsHinged lids are designed for easy one-handed opening, making it easier to access repeatedlyLids have a stacking feature for added stability and a secure locking tabLightweight lid design prevents tipping when open
Catalog No. NC3377676 Supplier CELLTREAT Scientific Products Supplier No. 229077
Empty LfTS Tip Racks available without a wafer to facilitate refills with LfTS Reload SystemsHinged lids are designed for easy one-handed opening, making it easier to access repeatedlyLids have a stacking feature for added stability and a secure locking tabLightweight lid design prevents tipping when open