Learn More
Waters Corp Waters Tip Seal Kit, XDS35i, XDS100B, Annual Replacement
Catalog No. 50792188 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Tip Seal Kit is a crucial component of the vacuum system, designed as a spare part for Scroll Pumps XDS35i and XDS100B. It ensures optimal performance and is recommended for annual replacement.
  • Part of the vacuum system
  • Used with Scroll Pump XDS35i
  • Used with Scroll Pump XDS100B
  • Recommended for annual replacement
  • One unit per package
Catalog No. 50-792-188 Supplier Waters Corp Supplier No. 6061146S
The Tip Seal Kit is a crucial component of the vacuum system, designed as a spare part for Scroll Pumps XDS35i and XDS100B. It ensures optimal performance and is recommended for annual replacement.
  • Part of the vacuum system
  • Used with Scroll Pump XDS35i
  • Used with Scroll Pump XDS100B
  • Recommended for annual replacement
  • One unit per package