Learn More
Andwin Scientific TIP SYRINGE 412 CURVED 500/CS
SDP
Catalog No. NC9619241
Encompass_Preferred
The curved tip syringe is ideal for pressure irrigation. This product has a volume capacity of 12 mL and has a reusable plastic finger grip extender. This is sold in a case of 500.
Catalog No. NC9619241 Supplier Andwin Scientific Supplier No. 8881412012CS
The curved tip syringe is ideal for pressure irrigation. This product has a volume capacity of 12 mL and has a reusable plastic finger grip extender. This is sold in a case of 500.