Learn More
Labcon North America Eclipse™ 250uL Pipet Tips, in Resealable Bags
Catalog No. 50101713
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits popular 250uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 1000 tips per bag; 10 bags per case; 52.1mm. H; Made in USA
Catalog No. 50-101-713 Supplier Labcon North America Supplier No. 10998000009
Tip, Pipet; Labcon; Eclipse™; Fits popular 250uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 1000 tips per bag; 10 bags per case; 52.1mm. H; Made in USA