Learn More
Labcon North America Eclipse™ 250uL Pipet Tips, in Racks
Catalog No. 50101714
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits popular 250uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 52.1mm. H; Made in USA
Catalog No. 50-101-714 Supplier Labcon North America Supplier No. 10999600089
Tip, Pipet; Labcon; Eclipse™; Fits popular 250uL pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 10 racks per pack; 10 packs per case; 52.1mm. H; Made in USA